Learn More
Cole-Parmer Great Plains Industries G2P10N09GMA Flowmeter/Totalizer, 5-50 GPM, 1" NPT(F), PVDF
Manufacturer: Cole-Parmer 0561012
This Flowmeter/Totalizer is ideal for use with many strong oxidizers and acids. It is battery-powered to eliminate the need for expensive wiring. It has two totalizers to allow for process or period-specific volume monitoring.