Learn More
Cole-Parmer Great Plains Industries A109GMA200NA2 Commercial Grade Flowmeter/Totalizer, 30-300 GPM, 2" NPT(F), Al  DFS Item
Manufacturer:  Cole-Parmer 0561008
A low-cost solution to light-duty process applications, this Flowmeter/Totalizer is battery-powered to eliminate the need for expensive wiring. It has two totalizers to allow for process or period-specific volume monitoring.
Catalog No. 50-631-044