Learn More
Cole-Parmer Masterflex Variable-Area Flowmeter, Correlated Reading, High-Flow, Aluminum Housing and Fittings, 65-mm Scale, 58.5 LPM Air  DFS Item
Manufacturer:  Cole-Parmer 0326640
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.
Catalog No. 50-627-261