Learn More
Dwyer Instruments Inc Flow Meter, 2"""" Scale, 30-240 CC/Min Air, SS Valve
SDP
Supplier:  Dwyer Instruments Inc RMA11SSV
Encompass_Preferred
Accurate within ±4% of full scale reading. Scales are brushed aluminum, coated with epoxy and the graduations are on both sides of the indicating tube.
Catalog No. NC0934166