Learn More
Cole-Parmer Great Plains Industries A109GMA025NA1 Flowmeter/Totalizer, 0.3-3 GPM, 1" NPT(F), Aluminum  DFS Item
Manufacturer:  Cole-Parmer 0561002
A low-cost solution to light-duty process applications, this Flowmeter/Totalizer is battery-powered to eliminate the need for expensive wiring. It has two totalizers to allow for process or period-specific volume monitoring.
Catalog No. 50-631-042