Learn More
Cole-Parmer Masterflex Variable-Area Flowtube, Correlated Reading, 150-mm Scale; 18.9 mL/min Air and 0.19 mL/min Water  DFS Item
Manufacturer:  Cole-Parmer 0321970
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs. Choose 65- or 150-mm flowtubes depending on the selected frame size. Flowtubes are calibrated for direct-reading or correlated reading of air or water at STP (standard temperature and pressure).
Catalog No. 50-627-149