Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Floxuridine, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
50-91-9
C9H11FN2O5
246.194
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
floxuridine,2'-deoxy-5-fluorouridine,5-fluoro-2'-deoxyuridine,5-fluorodeoxyuridine,fluorodeoxyuridine,floxuridin,fudr,deoxyfluorouridine,fluoruridine deoxyribose,fdurd
5790
CHEBI:60761
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
C1C(C(OC1N2C=C(C(=O)NC2=O)F)CO)O
This item is not returnable. View return policy
50-91-9
C9H11FN2O5
246.194
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
floxuridine,2'-deoxy-5-fluorouridine,5-fluoro-2'-deoxyuridine,5-fluorodeoxyuridine,fluorodeoxyuridine,floxuridin,fudr,deoxyfluorouridine,fluoruridine deoxyribose,fdurd
5790
CHEBI:60761
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
C1C(C(OC1N2C=C(C(=O)NC2=O)F)CO)O

Thymidylate synthetase inhibitor

Antineoplastic antimetabolite. Exhibits antiproliferative activity; inhibits thymidylate synthetase and disrupts DNA replication in human cells. Induces double-strand DNA breaks; activates ATR and ATM signaling pathways. Induces phosphorylation of Chk1 and Chk2 in OVCAR-8 and SKOV3ip ovarian cancer cell lines.
Quantity 50mg
Formula Weight 246.19
Percent Purity >98%
Chemical Name or Material Floxuridine

Recommended Storage : Store at Room Temperature