Learn More
Mouse anti-Flt-3/Flk-2/CD135, DyLight 550, Clone: 7E8.2C8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242210R
Catalog No. NBP242210R
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Flt-3/Flk-2/CD135
7E8.2C8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
P36888
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG2b Lambda
0.1 ml
Cell Cycle and Replication, Phospho Specific
Primary
2322
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Purified
FLT3
Partial recombinant human FLT3 protein (between amino acids 250-500) [UniProt P36888]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human