Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Mouse anti-Human, Cynomolgus Monkey, Alexa Fluor™ 532, Clone: MM0294-9G23, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212311AF532
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot.Specifications
Flt-3/Flk-2/CD135 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
FLT3 | |
Human recombinant Flt-3/Flk-2/CD135 | |
Primary | |
Human, Cynomolgus Monkey | |
IgG1 |
Western Blot, Flow Cytometry | |
MM0294-9G23 | |
Western Blot, Flow Cytometry | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
Mouse | |
0.1 mL | |
2322 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title