Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Mouse anti-Human, Cynomolgus Monkey, Alexa Fluor™ 594, Clone: MM0294-9G23, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212311AF594

Catalog No. NP212311A94


Add to cart

Description

Description

Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Flt-3/Flk-2/CD135
MM0294-9G23
Western Blot, Flow Cytometry
Conjugated, Purified
FLT3
Human Flt3/CD135
Protein G purified
Cell Cycle and Replication, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase
Primary
2322.0
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Cynomolgus Monkey, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit