Learn More
Flt-3/Flk-2/CD135 Mouse anti-Human, Cynomolgus Monkey, Alexa Fluor™ 750, Clone: MM0294-9G23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212311AF750
Catalog No. NP212311A70
Add to cart
Description
Description
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human,Cynomolgus Monkey samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
Flt-3/Flk-2/CD135
MM0294-9G23
Western Blot, Flow Cytometry
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Cynomolgus Monkey, Human
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FLT3
Human Flt3/CD135
0.1 mL
Primary
2322