Learn More
Flt-3/Flk-2/CD135 Mouse anti-Human, DyLight 488, Clone: 7B7C3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237363G
Catalog No. NBP237363G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Flt-3/Flk-2/CD135
7B7C3
Western Blot, ELISA, Immunohistochemistry
Purified
FLT3
Purified recombinant fragment of human FLT3 (AA: 930-991) expressed in E. Coli.
Protein A or G purified
Cell Cycle and Replication, Phospho Specific
Primary
2322
ELISA, Immunohistochemistry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human