Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Rat anti-Mouse, Alexa Fluor 405, Clone: 12B6, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11094155AF405

Catalog No. NBT94155405


Add to cart

Description

Description

Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Flt-3/Flk-2/CD135
12B6
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
FLT3
Recombinant Mouse Flt3/CD135 Antibody (12B6) extracellular domain
Protein G purified
Cell Cycle and Replication, Phospho Specific
Primary
2322
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes mouse Flt3/CD135 Antibody (12B6) in western blot.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit