Learn More
Alfa Aesar™ Flt-3 Inhibitor
Manufacturer:  Alfa Aesar™ J64484MF
 View more versions of this product
Catalog No. AAJ64484MF
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
Specifications
Specifications
50mg
2° to 8°C
A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
C18H20N2O4S
2-(3,4-Dimethoxybenzoylamino)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide
Signal Transduction Reagent
White
Powder
301305-73-7
360.43g/mol
Light Sensitive