Learn More
Flt-3 Ligand Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF308SP
Catalog No. AF308SP
Add to cart
This item is not returnable. View return policy
Description
Description
Flt-3 Ligand Polyclonal antibody AF308SP specifically detects Flt-3 Ligand in Human samples. It is validated for Neutralization, Western Blotting
Specifications
Specifications
Flt-3 Ligand
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.02-0.06 ug/mL
AAA17999
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Flt-3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Flt-3 is observed.
Human
Neutralization, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FLT3LG
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Flt-3 Ligand Thr27-Pro185 Accession # AAA17999
Antigen Affinity-purified
RUO
Primary
2323
Reconstitute at 0.2 mg/mL in sterile PBS.