Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3 Ligand Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI8D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270754AF647

Catalog No. N270754A647


Add to cart

Description

Description

Flt-3 Ligand Monoclonal antibody specifically detects Flt-3 Ligand in Human samples. It is validated for Western Blot.
Specifications

Specifications

Flt-3 Ligand
OTI8D10
Western Blot
Affinity Purified, Conjugated
FLT3LG
Recombinant protein expressed in E.coli corresponding to amino acids 27-181 of human Flt-3 Ligand.
Immunogen affinity purified
Biologically Active Proteins, Cell Cycle and Replication
Primary
2323.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit