Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Flt-3 Ligand Mouse Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $1298.00
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 2323 |
Molecular Weight | TMW: 19.3kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2521822
|
Novus Biologicals™
NBP2521820.02MG |
0.02mg |
Each for $344.00
|
|
NBP25218201
|
Novus Biologicals™
NBP2521820.1MG |
0.1mg |
Each for $1,298.00
|
|
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 27-189 of Mouse Flt-3 Ligand The Recombinant Mouse Flt-3 Ligand Protein is derived from Baculovirus. The Recombinant Mouse Flt-3 Ligand Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate buffered saline (pH 7.4) containing 10% glycerol | |
TMW: 19.3kDa | |
Baculovirus | |
FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine | |
Unlabeled |
SDS-Page | |
2323 | |
>85%, by SDS-PAGE | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Flt-3 Ligand | |
Yes |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title