Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLT4 Mouse anti-Human, Clone: 818CT12.1.1, Abnova™

Mouse monoclonal antibody raised against human FLT4.

Manufacturer:  Abnova Corporation MAB12341

Catalog No. 89-934-153


Add to cart

Description

Description

This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA. [provided by RefSeq]

Specifications

Specifications

FLT4
818CT12.1.1
Mouse monoclonal antibody raised against human FLT4.
In PBS (0.09% sodium azide)
FLT41/LMPH1A/PCL/VEGFR3
Mouse
IgG2a
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:25) Immunohistochemistry (1:25) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
FLT4
Recombinant His fusion protein corresponding to human FLT4.
Protein G purification
RUO
Primary
2324
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit