Learn More
MDI Membrane FLTR MDI NYLN 25MM 0.45 100PK
Supplier: MDI Membrane SYNN0602MNXX104
mdi Nylon-66 Syringe Filter, 25 mm, 0.45 µm, Inlet/Outlet: Female luer lock/male luer slip, HPLC Certified, non-sterile, 100 PK