Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flucloxacillin sodium, Alfa Aesar™

Click to view available options
Quantity:
100mg
1g
1847-24-1
C19H16ClFN3NaO5S
475.851
MFCD01682108
OTEANHMVDHZOPB-SLINCCQESA-M
101643208
CHEBI:31615
sodium;N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptan-6-yl]-3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboximidate
CC1=C(C(=NO1)C2=C(C=CC=C2Cl)F)C(=NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O)[O-].[Na+]
1847-24-1
C19H16ClFN3NaO5S
475.851
MFCD01682108
OTEANHMVDHZOPB-SLINCCQESA-M
101643208
CHEBI:31615
sodium;N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptan-6-yl]-3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboximidate
CC1=C(C(=NO1)C2=C(C=CC=C2Cl)F)C(=NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O)[O-].[Na+]
Applications
Flucloxacillin sodium is used to treat infections caused by susceptible Gram-positive bacteria. It is used in clinical in vitro tests known as antimicrobial susceptibility tests. It is used in treatment of pneumonia, bone infections, skin infections, wound infections and burns infections.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Solubility Information Soluble in water
Formula Weight 475.85
Physical Form Powder
Boiling Point 677°C
Color White
Melting Point 176°C to 178°C
Quantity 1g
Flash Point >110°C(230°F)
Sensitivity Light Sensitive
Chemical Name or Material Flucloxacillin sodium

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XH8680000