Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluconazole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
86386-73-4
C13H12F2N6O
306.28
RFHAOTPXVQNOHP-UHFFFAOYSA-N
2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F
86386-73-4
C13H12F2N6O
306.28
RFHAOTPXVQNOHP-UHFFFAOYSA-N
2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F

F1355, 86386-73-4, C13H12F2N6O

Spectrum™ Fluconazole, USP is used to treat and prevent superficial and systematic fungal infections. Known as a triazole antifungal drug, it has a white crystalline appearance and is slightly soluble in water and alcohol. Most commonly found under the trade names Diflucan and Trican and is listed on the World Health Organizations list of essential medicines. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 25 kg
CAS Max % 1
Percent Purity 98 to 102%