Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fludrocortisone Acetate, Micronized, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
514-36-3
C23H31FO6
422.49
SYWHXTATXSMDSB-GSLJADNHSA-N
2-[(1R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
514-36-3
C23H31FO6
422.49
SYWHXTATXSMDSB-GSLJADNHSA-N
2-[(1R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

FL117, 514-36-3, C23H31FO6

Spectrum™ Fludrocortisone Acetate, Micronized, USP is used along with other medications to treat low glucocorticoid levels caused by disease of the adrenal gland. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.001
Loss on Drying 0.03
Specific Rotation 126°-138°
Quantity 1 kg
CAS Max % 1
Percent Purity 97 to 103%