Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Flufenamic acid, 97%

Click to view available options
Quantity:
10g
50g
250g
530-78-9
C14H10F3NO2
281.234
MFCD00002422
LPEPZBJOKDYZAD-UHFFFAOYSA-N
flufenamic acid,fluphenamic acid,nichisedan,achless,arlef,flufacid,fullsafe,lanceat,paraflu,plostene
3371
CHEBI:42638
2-[3-(trifluoromethyl)anilino]benzoic acid
C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F
530-78-9
C14H10F3NO2
281.234
MFCD00002422
LPEPZBJOKDYZAD-UHFFFAOYSA-N
flufenamic acid,fluphenamic acid,nichisedan,achless,arlef,flufacid,fullsafe,lanceat,paraflu,plostene
3371
CHEBI:42638
2-[3-(trifluoromethyl)anilino]benzoic acid
C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F
Quantity 50g
Melting Point 133°C to 135°C
UN Number UN2811
Beilstein 1996069
Merck Index 14,4132
Formula Weight 281.23
Percent Purity 97%
Chemical Name or Material Flufenamic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CB4375000