Alfa Aesar™ Flufenamic acid, 97%

Manufacturer: Alfa Aesar™ B2358309

Quantity 10g

 View more versions of this product

Catalog No. AAB2358309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 530-78-9
    Molecular Formula C14H10F3NO2
    Formula Weight 281.23
    MDL Number MFCD00002422
    UN Number UN2811
    Beilstein 1996069
    Merck Index 14,4132
    Synonym N-(3-Trifluoromethylphenyl)anthranilic acid
    Chemical Name or Material Flufenamic acid

    Specifications

    Formula Weight 281.23
    Molecular Formula C14H10F3NO2
    MDL Number MFCD00002422
    UN Number UN2811
    Beilstein 1996069
    Merck Index 14,4132
    Synonym N-(3-Trifluoromethylphenyl)anthranilic acid
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number CB4375000
    Melting Point 133° to 135°C
    Quantity 10g
    CAS 530-78-9
    Chemical Name or Material Flufenamic acid
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit