Learn More
Honeywell™ Fluka™ Inert II SPE Columns
Inert plastic housing with PTFE frits
Supplier:  Honeywell™ 5028
 View more versions of this product
Catalog No. 600-29-46
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Reduced background contamination results in significant gains in detection capabilities
  • Decreases downtime for GC/MS source cleaning
  • Eliminates extraneous peaks
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 mL
40 to 63 μm
Yes
30
1000 mg
Cyclohexyl (CH)
60 Å
500 m2/g
Product Suggestions
Product Suggestions