Learn More
Honeywell™ Fluka™ Inert II SPE Columns
Inert plastic housing with PTFE frits
Supplier: Honeywell™ 5049
Description
- Reduced background contamination results in significant gains in detection capabilities
- Decreases downtime for GC/MS source cleaning
- Eliminates extraneous peaks
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
Specifications
25 mL | |
40 to 63 μm | |
60 Å |
5000 mg | |
Silica | |
50 |