Learn More
Honeywell™ Fluka™ Inert II SPE Columns
Inert plastic housing with PTFE frits
Manufacturer:  Honeywell™ 5008
 View more versions of this product
Catalog No. 600-30-52
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Reduced background contamination results in significant gains in detection capabilities
  • Decreases downtime for GC/MS source cleaning
  • Eliminates extraneous peaks
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
Specifications
Specifications
40 to 63 µm
1000 mg
Yes
30
6 mL
60 Å
500 m2/g
Octadecyl (C18)