Learn More
Honeywell™ Fluka™ Inert II SPE Columns
Inert plastic housing with PTFE frits
Products 16
Catalog Number Mfr. No. Sorbent Bed Weight Particle Size Price Quantity & Availability  
Catalog Number Mfr. No. Sorbent Bed Weight Particle Size Price Quantity & Availability  
600-29-03
View Documents
Honeywell™
5133
Alumina Neutral 3000 mg 50 to 200 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-29-46
View Documents
Honeywell™
5028
Cyclohexyl (CH) 1000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-29-49
View Documents
Honeywell™
5044
Diol 500 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-29-50
View Documents
Honeywell™
5048
Diol 1000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-29-79
View Documents
Honeywell™
5108
Florisil-PR 1000 mg 150 to 250 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-29-80
View Documents
Honeywell™
5109
Florisil-PR 2000 mg 150 to 250 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-29-81
View Documents
Honeywell™
5104
Florisil-PR 500 mg 150 to 250 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-30-35
View Documents
Honeywell™
5049
Silica 5000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
600-30-52
View Documents
Honeywell™
5008
Octadecyl (C18) 1000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-30-53
View Documents
Honeywell™
5010
Octadecyl (C18) 1500 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-30-54
View Documents
Honeywell™
5004
Octadecyl (C18) 500 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-30-55
View Documents
Honeywell™
5018
Octyl (C8) 1000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
600-30-56
View Documents
Honeywell™
5014
Octyl (C8) 500 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
60-031-07
View Documents
Honeywell™
5058
Silica 1000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
60-031-08
View Documents
Honeywell™
5059
Silica 2000 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
60-031-10
View Documents
Honeywell™
5054
Silica 500 mg 40 to 63 µm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Reduced background contamination results in significant gains in detection capabilities
  • Decreases downtime for GC/MS source cleaning
  • Eliminates extraneous peaks
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility