Learn More
Honeywell™ Fluka™ Inert II SPE, Empty Columns
Inert plastic housing with PTFE frits
Catalog Number | Mfr. No. | Column Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Column Capacity (Metric) | Description | Price | Quantity & Availability | ||||
600-28-89
|
Honeywell™
5435 |
8 mL | Bulk | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-30-41
|
Honeywell™
5444 |
25 mL | Loose frits | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-30-42
|
Honeywell™
5430 |
8 mL | No Frits | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
600-30-43
|
Honeywell™
5440 |
8 mL | Loose frits |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
- Reduced background contamination results in significant gains in detection capabilities
- Decreases downtime for GC/MS source cleaning
- Eliminates extraneous peaks
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
Specifications
Empty Solid Phase Extraction Column |