Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flumazenil, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
78755-81-4
C15H14FN3O3
303.29
OFBIFZUFASYYRE-UHFFFAOYSA-N
ethyl 12-fluoro-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
CCOC(=O)C1=C2CN(C)C(=O)C3=CC(F)=CC=C3N2C=N1
78755-81-4
C15H14FN3O3
303.29
OFBIFZUFASYYRE-UHFFFAOYSA-N
ethyl 12-fluoro-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
CCOC(=O)C1=C2CN(C)C(=O)C3=CC(F)=CC=C3N2C=N1

F3022, 78755-81-4, C15H14FN3O3

Spectrum™ Flumazenil, USP is used as an antidote in the treatment of benzodiazepine overdoses. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 1 g
CAS Max % 1
Percent Purity 98 to 102%