Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluocinolone Acetonide, Anhydrous, Micronized, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
250 g
Packaging:
Amber Glass Bottle
67-73-2
C24H30F2O6
452.50
FEBLZLNTKCEFIT-VSXGLTOVSA-N
(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
67-73-2
C24H30F2O6
452.50
FEBLZLNTKCEFIT-VSXGLTOVSA-N
(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

F1118, 67-73-2, C24H30F2O6

Spectrum™ Fluocinolone Acetonide, Anhydrous, Micronized, USP is a corticosteroid primarily used in dermatology to reduce skin inflammation and relieve itching. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Specific Rotation +98° to +108°
Loss on Drying 0.01
Quantity 5 g
CAS Max % 1
Percent Purity 97 to 102%