Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluor™escein Rabbit anti-Non-species specific, Alexa Fluor™ 350, Clone: 4-4-20 (enhanced), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252638AF350

Catalog No. NP22638A30


Add to cart

Description

Description

Fluorescein Monoclonal antibody specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Fluorescein
4-4-20 (enhanced)
Western Blot, Flow Cytometry
Conjugated, Purified
Rabbit
IgG Kappa
0.1 mL
Primary
Fluorescein labelled proteins.
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FITC, fluorescein isothyocianate
Fluorescein I-bovine serum albumin (FlI-BSA).
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Nonspecies specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit