Learn More
BioVision IncSupplier Diversity Partner CaspGLOW™ Fluorescein Active Caspase-8 Staining Kit: Convenient & Sensitive Kit to Detect Activated Caspases-8 in Living Cells. Detection Method: Fluorescence Microscopy, Flow Cytometry or Fluorescence Plate Reader. Kit Size: 25 and 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K18825
Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Active Caspase-8 Staining Kit provides a convenient means for sensitive detection of activated caspase-8 in living cells. The assay utilizes the caspase-8 inhibitor, IETD-FMK, conjugated to FITC (FITC-IETD-FMK) as a fluorescent in situ marker. FITC-IETD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-8 in apoptotic cells. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9594