Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescamine, Alfa Aesar™

Click to view available options
Quantity:
25mg
100mg
500mg
38183-12-9
C17H10O4
278.263
MFCD00005928
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
fluorescamine,fluram,spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl,4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione,4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione,4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione,zfkjvjidpqddfy-uhfffaoysa,4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
37927
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3
38183-12-9
C17H10O4
278.263
MFCD00005928
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
fluorescamine,fluram,spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl,4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione,4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione,4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione,zfkjvjidpqddfy-uhfffaoysa,4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
37927
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3

For fluorescent detection of amino acids, peptides and proteins

Color White to Cream
Quantity 25mg
Melting Point 155°C to 158°C
Merck Index 14,4158
Solubility Information Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water.
Formula Weight 278.27
Physical Form Powder
Chemical Name or Material Fluorescamine

EINECSNumber : 253-814-5

TSCA : No

Recommended Storage : Ambient temperatures