Learn More
Aat BioquestSupplier Diversity Partner Fluorescamine *CAS 38183-12-9* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 820
Fluorescamine is a popular amine-fluorogenic reagent for determining protein concentrations in solutions and on gels. It is also used to analyze low molecular weight amines by TLC, HPLC and capillary electrophoresis.
Catalog No. 50-195-2951