Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescamine, pure, ACROS Organics™

Click to view available options
Quantity:
25mg
100mg
500mg
2.5g
Packaging:
Glass bottle
38183-12-9
C17H10O4
278.263
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
37927
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3
38183-12-9
C17H10O4
278.263
ZFKJVJIDPQDDFY-UHFFFAOYSA-N
37927
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
C1=CC=C(C=C1)C2=COC3(C2=O)C4=CC=CC=C4C(=O)O3
Additional Information Fluorescence ind. for prim. amines and amino acids
Infrared Spectrum Authentic
Formula Weight 278.26
Physical Form Powder
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: .. Other solubilities: soluble in diethyl ether
Specific Extinction 900 (235nm, ether) min.
Melting Point 155.0°C to 157.0°C
Color Yellow
Quantity 2.5g
Chemical Name or Material Fluorescamine
Show More Show Less