Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein, Alcohol Soluble, Spectrum™

Click to view available options
Quantity:
25 g
100 g
2.5 kg
Packaging:
Amber Glass Bottle
2321-07-5
2321-07-5
CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 25 g

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.