Learn More
Molecular Probes™ Fluorescein Cadaverine; 5-((5-Aminopentyl)thioureidyl)Fluorescein, Dihydrobromide Salt Non-distribution product as customer accommodation.
5-((5-aminopentyl)thioureidyl)fluorescein, dihydrobromide salt (fluorescein cadaverine)
Manufacturer:  Molecular Probes™ A10466
Catalog No. A10466
Add to cart
Description
Description
The primary aliphatic amine of fluorescein cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
Specifications
Specifications
Fluorescein Cadaverine; 5-((5-Aminopentyl)thioureidyl)Fluorescein, Dihydrobromide Salt, 25mg, Yellow, Solid, C26H27Br2N3O5S Molecular Formula, 653.38 Molecular Weight, FITC (Fluorescein) Label, Aldehyde, Carboxylic Acid, Ketone Chemical Reactivity, 493/51
Yellow
FITC (Fluorescein)
Room temperature
653.38kDa
517nm
Solid
Labeling chemistry, protein labeling and crosslinking
Fluorescein Cadaverine; 5-((5-Aminopentyl)thioureidyl)Fluorescein, Dihydrobromide Salt
25mg
493nm
HPLC, 99% at 254nm
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.