Learn More
Fluorescein, Clone: 4-4-20 (enhanced), Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00203
Catalog No. 89-032-544
Add to cart
Specifications
Specifications
Fluorescein
4-4-20 (enhanced)
Flow Cytometry Negative Control Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG4, kappa
RUO
Antibody
Monoclonal
Flow Cytometry, Western Blot
Unconjugated
In PBS (0.02% Proclin 300).
Original antibody is raised against fluorescein I-bovine serum albumin (FlI-BSA).
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
Fluorescein labelled proteins.