Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fluorescein, Clone: 4-4-20 (enhanced), Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00079
Catalog No. 89-032-422
Specifications
Specifications
Fluorescein
4-4-20 (enhanced)
Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG, kappa
RUO
Antibody
Monoclonal
Flow Cytometry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against Fluorescein I conjugated with BSA.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
This antibody detects Fluorescein labelled proteins.