Alfa Aesar™ Fluorescein diacetate, 97%

Manufacturer: Alfa Aesar™ B2446622

Quantity 100g

 View more versions of this product

Catalog No. AAB2446622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 596-09-8
    Molecular Formula C24H16O7
    Formula Weight 416.39
    MDL Number MFCD00005062
    Beilstein 62575
    Chemical Name or Material Fluorescein diacetate

    Specifications

    Formula Weight 416.39
    Assay Percent Range 0.97
    TSCA TSCA
    Molecular Formula C24H16O7
    MDL Number MFCD00005062
    CAS 596-09-8
    Beilstein 62575
    Melting Point 200° to 203°C
    Quantity 100g
    Chemical Name or Material Fluorescein diacetate
    Assay 97%

    Description

    Applications
    Fluorescein diacetate stains living cells green, and is a substrate for esterases. It is suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes. It is used in studies on intracellular interactions and membrane permeability.

    Solubility
    Soluble in chloroform, DMSO, ethanol, and water.

    Notes
    Store at -20°C. Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit