Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein diacetate, 97%

Manufacturer:  Alfa Aesar B2446606

Quantity 5g

 View more versions of this product

Catalog No. AAB2446606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Fluorescein diacetate stains living cells green, and is a substrate for esterases. It is suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes. It is used in studies on intracellular interactions and membrane permeability.

Solubility
Soluble in chloroform, DMSO, ethanol, and water.

Notes
Store at -20°C. Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

596-09-8
416.39
62575
C24H16O7
MFCD00005062
Fluorescein diacetate

Specifications

596-09-8
C24H16O7
MFCD00005062
5g
97%
0.97
416.39
62575
Fluorescein diacetate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit