Learn More
Aat BioquestSupplier Diversity Partner Fluorescein, disodium salt *CAS 518-47-8* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 2
Fluorescein, disodium salt is water-soluble version of fluorescein (see product #1). Fluorescein is a fluorophore widely used in microscopy, forensics and serology to detect latent blood stains.
Catalog No. 50-195-2676