Learn More
Fluorescein-Ubiquitin
Fluorescein-Ubiquitin
$340.00 - $1454.00
Specifications
Conjugate | Fluorescein |
---|---|
Content And Storage | Fluorescein-labeled ubiquitin in HEPES buffered saline. Store at -20°C. |
Detection Method | Fluorescent |
Excitation/Emission | 340⁄490, 520 |
For Use With (Equipment) | Microplate Reader |
Description
Fluorescein-labeled ubiquitin for use as a general research tool or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.For use in combination with FRET donors including LanthaScreen™ Tb-Ubiquitin (PV4375) or a LanthaScreen™ antibody including Tb-anti-GST (PV4216) or Tb-anti-ubiquitin-FK2 (PV4751).
Specifications
Fluorescein | |
Fluorescent | |
Microplate Reader | |
Classic Dyes | |
Dry Ice | |
TR-FRET |
Fluorescein-labeled ubiquitin in HEPES buffered saline. Store at -20°C. | |
340⁄490, 520 | |
FITC (Fluorescein) | |
End Point | |
Protein-Based Substrate | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.