Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein, Water Soluble, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
518-47-8
518-47-8
CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 25 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.