Learn More
Molecular Probes™ Fluoresceinyl Glycine Amide (5-(Aminoacetamido)Fluorescein) Non-distribution product as customer accommodation.
Fluoresceinyl Glycine Amide (5-(Aminoacetamido)Fluorescein)
Manufacturer:  Molecular Probes™ A1363
Catalog No. A1363
Add to cart
Description
Description
The primary aliphatic amine of 5-(aminoacetamido)fluorescein (fluoresceinyl glycine amide) can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). The glycine may be the better amine probe for direct carbodiimide-mediated coupling, since it is likely to remain unprotonated at a lower pH than other aliphatic amines.
Specifications
Specifications
Fluoresceinyl Glycine Amide (5-(Aminoacetamido)Fluorescein), 10mg, Orange, Solid, C22H16N2O6 Molecular Formula, 404.38 Molecular Weight, FITC (Fluorescein) Label, Aldehyde, Carboxylic Acid, Ketone Chemical Reactivity, 515/491nm Excitation/Emission Wavelen
Orange
FITC (Fluorescein)
Room temperature
404.38kDa
515nm
Solid
Labeling chemistry, protein labeling and crosslinking
Fluoresceinyl Glycine Amide (5-(Aminoacetamido)Fluorescein)
10mg
491nm
HPLC, 98% at 254nm
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.