Learn More
Motic Fluorescence Filter Set for BA410E/AE31E
Catalog No. 01257477 Shop All Motic Group Products
Change view
Click to view available options
Product Line:
DAPI/Hoechst/AlexaFluor 350
EGFP/FITC/Cy2/AlexaFluor 488
TRITC/Cy3/TagRFP/AlexaFluor 546
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
01-257-477 EGFP/FITC/Cy2/AlexaFluor 488
01-257-476 DAPI/Hoechst/AlexaFluor 350
01-257-478 TRITC/Cy3/TagRFP/AlexaFluor 546
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-257-477 Supplier Motic Supplier No. 1101000203281
Add to Cart
Add to Cart
This item is not returnable. View return policy
This is an accessory part for the BA410E series and AE31E series. Filter sets require an EPI-Fluorescence attachment for BA or attachment for AE to function.
    li>Valid for BA410E Binocular Sextuple 50W, BA410E Binocular Sextuple 100W, BA410E Trinocular Sextuple 50W, BA410E Trinocular Sextuple 100W, AE31E Binocular, AE31E Trinocular, AE31E Binocular 100W, AE31E Trinocular 100W
Specifications
Type Filter Cube
Product Line EGFP/FITC/Cy2/AlexaFluor 488
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov