Learn More
Abnova™ Fluorescent Exosome Standard (A549 cell line) (Green dye)
This product is purified exosomes obtained from A549 cell line (human lung carcinoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Manufacturer:  Abnova™ U0373
Catalog No. 89-998-455
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.