Learn More
Labconco™ Fluorescent Light Kit
SDPSCI_ED
Labconco Fluorescent Light Kit provides interior lighting for Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and XPert™ Enclosures.
Supplier:  Labconco™ 3909200/EMD
Catalog No. S28152
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • May be placed atop enclosure to enhance illumination of interior
  • More than one may be used on larger enclosures
  • Auxiliary electrical receptacle with switch for connection of accessories such as printer or airflow monitor
  • Includes IEC, three-wire cord with plug
Warranty and Services
One year
Specifications
Specifications
Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and XPert™ enclosures
15 lb.
18.3 in.
Lamp and Power Cord with Plug
46.4 cm
60 Hz
3.0 in.
6.8 Kg
7.62 cm
10 A
115 V
Product Suggestions
Product Suggestions