Learn More
Bachem 5(6)-FAM-(Glu¹)-Orexin A (human, mouse, rat), 0.5mg, 1678416-64-2, Trifluoroacetate salt DFS Item
Manufacturer:  Bachem 4091433.0500
Fluorescently labeled orexin A, absorption at 495 nm, emission at 519 nm.
Catalog No. 50-194-6964